Learn More
Invitrogen™ POU4F1 Recombinant Rabbit Monoclonal Antibody (9A5O9)
Rabbit Recombinant Monoclonal Antibody
$169.00 - $419.00
Specifications
Antigen | POU4F1 |
---|---|
Clone | 9A5O9 |
Concentration | 1.04 mg/mL |
Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
Applications | ELISA, Western Blot |
Description
Immunogen sequence: MMSMNSKQPH FAMHPTLPEH KYPSLHSSSE AIRRACLPTP PLQSNLFASL DETLLARAEA LAAVDIAVSQ GKSHPFKPDA TYHTMNSVPC TSTSTVPLAH.
BRN3A is a class IV POU domain-containing transcription factor highly expressed in the developing sensory nervous system and in cells of the B- and T-lymphocytic lineages.Specifications
POU4F1 | |
1.04 mg/mL | |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
Q01851 | |
5457 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human POU4F1 (NP_006228.3). | |
Antibody |
9A5O9 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human | |
Pou4f1 | |
Brain-3A; brain-specific homeobox/POU domain protein 3A; Brn3; Brn-3; brn3.0; Brn-3.0; Brn3a; brn-3A; brn3b; E130119J07Rik; etID22526.1; FLJ13449; homeobox/POU domain protein RDC-1; LOW QUALITY PROTEIN: POU domain, class 4, transcription factor 1; Oct-T1; POU class 4 homeobox 1; POU domain, class 4, transcription factor 1; POU4F1; pou4f1 protein; RDC1; RDC-1 | |
Pou4f1 | |
Primary | |
Protein A |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.