Learn More
Aviva Systems Biology Corp POU5F2 ANTIBODY 100UG/100UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP39945_T100
This is a rabbit polyclonal antibody against POU5F2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human POU5F2. Alternate names: DKFZp686P02123, FLJ25680, SPRM-1ApplicationWB. Species reactivity: Human, Mouse
Catalog No. 50-014-53166