Learn More
Cole-Parmer AEMC 8220 W/MN193-BK Power Quality Meter with 6A/120Aac Probe Model 8220 with MN93-Bk  DFS Item
Manufacturer:  Cole-Parmer 2001982
This current probe measures up to 600 Vrms or VDC, and up to 6500 AAC or 1400 ADC, which is probe dependent. It displays the minimum, maximum and average volts and amps, crest factor, peak value and K-factor. The powerpad calculates and displays watts, VARs and VA, power factor and displacement power factor for single-phase and balanced three-phase. It also displays total harmonic distortion for voltage and current, as well as individual harmonic values and percentage for volts and amps through the 50 harmonic, and captures and displays in rush current. In addition, it calculates and displays phase rotation and RPM, as well as temperature in both F and C, resistance up to 2,000, and conducts continuity and diode tests. The probe stores up to nine complete sets of readings for all volt, amp, power and harmonic measurements. It is configurable from DataView software via optically coupled RS232 port and operates off batteries or a recharger.
Catalog No. 50-627-348