Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SW Safety PowerChem PC-115GR 5.9 mil Extended-Cuff Neoprene Gloves
SDP

This heavy-duty latex-free neoprene glove provides superior resistance to chemicals, punctures and abrasions with excellent grip, optimum tactility and an extended cuff for splash protection.

$14.93

Specifications

ANSI Abrasion Rating ANSI 0
Certifications/Compliance FDA 21 CFR 177.2600, ASTM D6977-19, ASTM F1671-13, ASTM F739-12
Certifications/Compliance (Europe) EN ISO 21420:2020, EN 455-2000-2015, Reach EU 1907/2006, EU 2016/425
Applications Healthcare, Laboratory
Color Green
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-441-074
SDP
View Documents
SW Safety
K071402
Small
Pack of 50 for $14.93
Case of 10 PK for $149.30
Add to cart
 
17-441-075
SDP
View Documents
SW Safety
K071403
Medium
Pack of 50 for $14.93
Case of 10 PK for $149.25
Add to cart
 
17-441-076
SDP
View Documents
SW Safety
K071404
Large
Pack of 50 for $14.93
Case of 10 PK for $149.25
Add to cart
 
17-441-077
SDP
View Documents
SW Safety
K071405
X-Large
Pack of 50 for $14.93
Case of 10 PK for $149.25
Add to cart
 
17-441-078
SDP
View Documents
SW Safety
K071406
2X-Large
Pack of 50 for $14.93
Case of 10 PK for $149.25
Add to cart
 
Description

Description

  • Pure neoprene prevents latex allergic reactions
  • Extended cuff for added protection
  • Textured surface delivers excellent grip
  • Ergonomic design for comfort
  • Optimal thickness for durability and tactile sensitivity
  • Powder-free to prevent contamination
  • Tested for use with chemotherapy drugs
  • SHA Dermatologically Approved
  • 1.5 AQL for wet and dry environments
Specifications

Specifications

ANSI 0
EN ISO 21420:2020, EN 455-2000-2015, Reach EU 1907/2006, EU 2016/425
Green
7.5 mil
Yes
5.9 mil
No
Beaded
PC-115 GR
11.5 in.
FDA 21 CFR 177.2600, ASTM D6977-19, ASTM F1671-13, ASTM F739-12
Healthcare, Laboratory
Partially Textured
0.19 mm
Neoprene
0.15 mm
Type C
Unlined
50 Pk., 500/Cs.
29 cm
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit