Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SW Safety PowerForm PF-95GY 6.2 mil Biodegradable Sweat-Absorbing Nitrile Gloves with DriTek and EcoTek
GREENER_CHOICESDP

Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large

Biodegradable nitrile gloves allow workers to safely perform tasks, while remaining cool and comfortable with exclusive DriTek Sweat Management Technology. Fully textured surface enhances grip capabilities in wet and dry conditions.

  • DriTek Comfort Technology with absorbent lining that wicks away moisture for added extended use
  • EcoTek Biodegradability Technology accelerates landfill breakdown without any performance loss
  • Fully textured for secure grip
  • Silicone-free for critical environments
  • 1.5 AQL exceeds laboratory standards
  • ANSI 3 Abrasion resistance
  • ANSI 1 Puncture resistance
ANSI Abrasion Rating ANSI 3
Certifications/Compliance ASTM D6319-19, ASTM D6978-05 (2019), ASTM F1671-13, ASTM F739-12, FDA 21 CFR 177.2600
Certifications/Compliance (Europe) EN ISO 21420:2020, EN 455-2000-2015, Reach EU 1907/2006, EU 2016/425
Chemotherapeutic Rated No
Applications Industrial, Laboratory
AQL 1.5
Color Gray
Disposable Yes
Exterior Finish Fully Textured
Finger Thickness (English) 8.2 mil
Finger Thickness (Metric) 0.21 mm
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Palm Thickness (English) 6.2 mil
Palm Thickness (Metric) 0.16 mm
Powdered Powder Free
Size X-Large
Thickness (English) 8.2 mil
Thickness (Metric) 0.21 mm
Series PF-95GY
Show More Show Less

Product tested under ASTM D5526-12, a standard test method for determining anaerobic biodegradation of plastics in accelerated landfill conditions. Current biodegradation results do not predict future results.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit