Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SW™ PowerForm™ Gray Nitrile Gloves with DriTek™ and EcoTek™

Biodegradable nitrile gloves allow workers to safely perform tasks, while remaining cool and comfortable with exclusive DriTek™ Sweat Management Technology. Fully textured surface enhances grip capabilities in wet and dry conditions.

$286.82

Specifications

Applications Industrial, Laboratory
Certifications/Compliance FDA 21 CFR 177.2600
Glove Cuff Style Beaded
AQL 1.5
Color Gray
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-441-085
View Documents
SW™
N260501
X-Small
Case of 500 for $286.82
Add to cart
 
17-441-086
View Documents
SW™
N260502
Small
Case of 500 for $286.82
Add to cart
 
17-441-087
View Documents
SW™
N260503
Medium
Case of 500 for $286.82
Add to cart
 
17-441-089
View Documents
SW™
N260505
X-Large
Case of 500 for $286.82
Add to cart
 
17-441-090
View Documents
SW™
N260506
2X-Large
Case of 500 for $286.82
Add to cart
 
Description

Description

  • EcoTek™ Biodegradability Technology accelerates landfill breakdown without any performance loss
  • DriTek™ Comfort Technology with absorbent lining that wicks away moisture for added extended use
  • Fully textured for enhanced grip in wet and dry conditions
  • Optimal thickness for durability and tactile sensitivity
  • Silicone-free for critical environments
  • Touch-screen compatible
  • 1.5 AQL exceeds laboratory standards
  • SHA Dermatologically Approved
Specifications

Specifications

Industrial, Laboratory
Beaded
Gray
Fully Textured
Yes
6.2 mil
PF-95GY
FDA 21 CFR 177.2600
1.5
Yes
8.2 mil
Nitrile
Powder Free
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit