Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wearwell Powerhouse™ 5 Channel Straight Cable Protectors

Secure Lid-Lock feature protects pedestrian traffic by preventing trip hazards. Bright yellow lid is highly visible for enhanced safety and quick recognition.

$293.00 - $366.00

Specifications

Color Black/Yellow
For Use With (Application) Cable Protection
Length (Metric) 91 cm
Material Natural Rubber
Size (English) 36 x 19.75 x 3 in.
View More Specs

Products 3
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
19-003-161
View Documents
Wearwell
PWR.5CHAN.GP.BYL
N/A General Purpose
Each for $326.00
Add to cart
 
19-003-163
View Documents
Wearwell
PWR.5CHAN.MD.BYL
N/A Medium Duty
Each for $293.00
Add to cart
 
19-003-162
View Documents
Wearwell
PWR.5CHAN.HD.BYL
N/A Heavy Duty
Each for $366.00
Add to cart
 
Description

Description

  • Highly resistant to corrosion from oils and chemicals
  • Guards against impact and abrasion
  • Hidden hinges allow the lid to lie flat while protecting cables from damage
  • Hidden interlocks keep adjoining sections from lifting
  • Lid features a built-in access port for easy opening with a screwdriver or pry bar
  • Built-in underside handle makes relocating and handling easy
  • Secure Lid-Lock feature protects pedestrian traffic by preventing trip hazards
  • Available in General Purpose, Medium Duty, and Heavy Duty options
Specifications

Specifications

Black/Yellow
91 cm
36 x 19.75 x 3 in.
12 cm
Cable Protection
Natural Rubber
3 in.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit