Learn More
Cole-Parmer AEMC 8230 W/MN93-BK Powerpad Jr. 8230 with 240A Current Probe Model 8230 with MN93-Bk  DFS Item
Manufacturer:  Cole-Parmer 2001971
Measures up to 600 Vrms or VDC
Measures up to 6500 AAC or 1400 ADC (probe dependent)
Displays min, max and average volts and amps, crest factor, peak value and K-factor
Calculates and displays watts, VARs and VA, Power FActor and displacement power factor for single-phase and balanced three-phase
Displays total harmonic distortion (THD-F and THD-R) for voltage and current
Displays individual harmonic values and % for volts and amps through the 50th harmonic
Captures up to 4096 alarm events from up to 10 different thresholds
Displays and records up to 17 different power quality parameters
Standard accessories include: current probes with 10 ft leads and black connector, black and red 10 ft voltage leads and alligator clips, RS232 DB9F optically coupled serial cable, NiMH battery, US 120V power cord, DataView software, carrying bag, soft carrying pouch and user manual
Catalog No. 50-627-337