Learn More
Andwin ScientificSupplier Diversity Partner PP TUBE 12X75 5ML YL 1000/CS DFS Item
Manufacturer:  Andwin ScientificSupplier Diversity Partner 60818432
The Culture Tubes are constructed with polypropylene. This product is autoclavable, disposable, and nonsterile. The tubes have a volume capacity of 5 mL, an outer diameter of 12 mm, and length of 75 mm. This is sold in a case of 1000 yellow tubes.
Catalog No. NC0207427