Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PP1 alpha/PPP1A Mouse anti-Human, Mouse, Rat, Clone: OTI6E5, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273542
Description
PP1 alpha/PPP1A Monoclonal specifically detects PP1 alpha/PPP1A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PP1 alpha/PPP1A | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
EC 3.1.3.16, PP1alpha, PP-1AMGC15877, PPP1AMGC1674, protein phosphatase 1, catalytic subunit, alpha isoform, protein phosphatase 1, catalytic subunit, alpha isozyme, serine/threonine protein phosphatase PP1-alpha 1 catalytic subunit, serine/threonine-protein phosphatase PP1-alpha catalytic subunit | |
Mouse | |
38.5 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI6E5 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
PPP1CA | |
Full length human recombinant protein of human PPP1CA(NP_001008709) produced in E.coli. | |
Affinity Purified | |
Phospho Specific, Protein Phosphatase | |
5499 | |
Human, Mouse, Rat | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title