Learn More
Invitrogen™ PP1 beta Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578117
Description
Positive Control: 293T, A431, HeLa, HepG2, mouse midbrain, rat brain Predicted Reactivity: Zebrafish (97%), Xenopus laevis (100%), Dog (98%), Pig (100%), Rabbit (100%), Chicken (100%), Rhesus Monkey (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is one of the three catalytic subunits of protein phosphatase 1 (PP1). PP1 is a serine/threonine specific protein phosphatase known to be involved in the regulation of a variety of cellular processes, such as cell division, glycogen metabolism, muscle contractility, protein synthesis, and HIV-1 viral transcription. Mouse studies suggest that PP1 functions as a suppressor of learning and memory. Two alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications
PP1 beta | |
Polyclonal | |
Unconjugated | |
Ppp1cb | |
1200010B19; epididymis secretory sperm binding protein Li 80p; HEL-S-80p; hypothetical protein LOC538829; MGC21657; PP1B; PP-1B; PP1beta; PP1-BETA; PP2CB; PP2CBETA; PP2C-beta-X; PPC2BETAX; PPP1CB; PPP1CD; protein phosphatase 1; protein phosphatase 1 catalytic subunit beta; protein phosphatase 1, catalytic subunit, beta isoform; protein phosphatase 1, catalytic subunit, beta isozyme; protein phosphatase 1, catalytic subunit, delta isoform; protein phosphatase 1-beta; protein phosphatase 1-delta; protein phosphatase-1a; serine/threonine protein phosphatase PP1-beta catalytic subunit; serine/threonine-protein phosphatase PP1-beta catalytic subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19046, 25594, 5500 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1.0 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
P62140, P62141, P62142 | |
Ppp1cb | |
Recombinant protein encompassing a sequence within the center region of human PPP1CB. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.