Learn More
PP5 Mouse anti-Human, Alexa Fluor 405, Clone: 12F7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP259308AF405
Catalog No. NB259308405
Add to cart
Description
Description
PP5 Monoclonal specifically detects PP5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
PP5
12F7
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
PPP5C
Full length human PP5 protein
Protein G purified
Cell Cycle and Replication
Primary
5536
Human
Western Blot, Immunocytochemistry, Immunofluorescence
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
EC 3.1.3.16, PP5FLJ36922, PPP5, PPT, PP-T, protein phosphatase 5, catalytic subunit, Protein phosphatase T, Serine/Threonine phosphatase, serine/threonine-protein phosphatase 5
Mouse
IgG1
100 ul
Store at 4C in the dark.
Monoclonal
Detects 58kDa.