Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PP5 Antibody (2E11), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259307UV
Description
PP5 Monoclonal specifically detects PP5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
PP5 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PPP5C | |
Full length human PP5 protein | |
100 μL | |
Primary | |
Detects 58kDa. | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, Immunocytochemistry, Immunofluorescence | |
200000000000 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
EC 3.1.3.16, PP5FLJ36922, PPP5, PPT, PP-T, protein phosphatase 5, catalytic subunit, Protein phosphatase T, Serine/Threonine phosphatase, serine/threonine-protein phosphatase 5 | |
Mouse | |
Protein G purified | |
Cell Cycle and Replication | |
5536 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction