Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPAP2A Mouse anti-Human, Clone: OTI1H4, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273564
Description
PPAP2A Monoclonal specifically detects PPAP2A in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PPAP2A | |
Monoclonal | |
LYOPH | |
Western Blot 1:500, Immunohistochemistry 1:150 | |
EC 3.1.3.4, lipid phosphate phosphohydrolase 1, lipid phosphate phosphohydrolase 1a, LLP1a, LPP1PAP2a2, PAP2, PAP2a, PAP2-alpha, PAP-2aPAP2alpha2, PAPalpha1, Phosphatidate phosphohydrolase type 2a, Phosphatidic acid phosphatase 2a, phosphatidic acid phosphatase type 2A, phosphatidic acid phosphohydrolase type 2a, type 2 phosphatidic acid phosphohydrolase, type-2 phosphatidic acid phosphatase alpha | |
Mouse | |
32 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1H4 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
PLPP1 | |
Full length human recombinant protein of human PPAP2A(NP_003702) produced in HEK293T cell. | |
Affinity Purified | |
Cell Cycle and Replication | |
8611 | |
Human | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title