Learn More
PPAPDC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 206351AP
Catalog No. 50-173-3004
Add to cart
Description
Description
PPAPDC3, also known as nuclear envelope transmembrane protein 39 (NET39), was initially discovered in an in silico screen for secreted or membrane proteins. It is a member of the PAP2 superfamily of phosphatases and haloperoxidases. PPAPDC3 has recently been shown to act as a negative regulator of myoblast differentiation by diminishing the activity of the mammalian target of rapamycin TOR. PPAPDC3 is highly expressed in cardiac and skeletal muscle and becomes strongly upregulated during cultured myoblast differentiation tissues. Overexpression of PPAPDC3 in myoblasts repressed myogenesis while knockdown by RNA interference promoted differentiation indicating its part in the regulatory mechanism for myogenesis.
Specifications
Specifications
PPAPDC3
0.49 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5FVJ3, Q8NBV4, Q91WB2
C9orf67, KIAA0515, NET39, PPAPDC3
Rabbit
IgG
150 μL
-20°C
Primary
227721, 296635, 84814
Immunoprecipitation, Western Blot
Unconjugated
PLPP7
Liquid
Plpp7
PPAPDC3 Fusion Protein Ag14676
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat