Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPAR delta/NR1C2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen PPAR delta/NR1C2
Applications Immunofluorescence, KnockDown, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093576
View Documents
Novus Biologicals
NBP303214100UL
100 μg Each for $320.71
Add to cart
 
NB093577
View Documents
Novus Biologicals
NBP30321420UL
20 μg Each for $138.37
Add to cart
 
Description

Description

PPAR delta/NR1C2 Polyclonal antibody specifically detects PPAR delta/NR1C2 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence, KnockDown
Specifications

Specifications

PPAR delta/NR1C2
Unconjugated
Rabbit
Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators, Wnt Signaling Pathway
PBS with 50% glycerol, pH7.3.
5467
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence, KnockDown, Western Blot
Purified
RUO
Human, Mouse
FAAR, MGC3931, NR1C2PPAR-delta, NUC1PPAR-beta, NUCI, NUCII, Nuclear hormone receptor 1, Nuclear receptor subfamily 1 group C member 2, peroxisome proliferative activated receptor, delta, Peroxisome proliferator-activated receptor beta, peroxisome proliferator-activated receptor delta, PPARB
Recombinant fusion protein containing a sequence corresponding to amino acids 102-361 of human PPAR delta/NR1C2 (NP_803184.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit