Learn More
PPAR gamma/NR1C3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259222
Catalog No. NBP259222
Add to cart
Description
Description
PPAR gamma/NR1C3 Polyclonal antibody specifically detects PPAR gamma/NR1C3 in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
PPAR gamma/NR1C3
Unconjugated
Serum with 0.05% Sodium Azide
CIMT1, NR1C3GLM1, Nuclear receptor subfamily 1 group C member 3, peroxisome proliferator-activated receptor gamma, peroxisome proliferator-activated receptor gamma 1, PPAR gamma, PPARG1peroxisome proliferative activated receptor, gamma, PPARG2PPARgamma, PPAR-gamma
Rabbit
IgG
100 ul
Cancer, mTOR Pathway
Primary
5468
Western Blot, ELISA
Western Blot 1:500, ELISA 1:500
Antiserum
PPARG
The exact immunogen is propietary information.
Whole Antiserum
RUO
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Polyclonal
Human