Learn More
Invitrogen™ PPARA Recombinant Human Protein, Ligand Binding Domain, GST-Tagged
PPARA (peroxisome proliferator activated receptor alpha) ligand binding domain is a 59.40 kDa recombinant human protein (amino acids 192-468) expressed as a GST-tagged fusion protein in baculovirus-infected insect cells.
Supplier: Invitrogen™ PV4692

Description
PPAR alpha-LBD was expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator displacement studies.
Specifications
5465 | |
PPAR alpha | |
GST Tag | |
Dry Ice | |
Ligand Binding Domain, Recombinant | |
Hormone Receptors | |
Human |
50 µg | |
Yes | |
Store in ultra-cold freezer (-68 to -85°C). | |
Kinases & Inhibitors | |
Liquid | |
Ligand Binding Assay | |
Neurobiology |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.