Sino Biological™ PPARG Recombinant Human Protein, N-His.GST tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human PPARG isoform 2 (Met 1-Tyr 505) fused to N-terminal polyhistidine-tagged GST tag at N-Terminus

Manufacturer: Sino Biological™ 12019H20B5

 View more versions of this product

Catalog No. 12-019-H20B5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 85.4kd
    Purity ≥90% by SDS-PAGE
    Quantity 5μg
    Source Human
    Structural Form Full Length, Recombinant
    Endotoxin Concentration <1 EU/μg
    Common Name Peroxisome Proliferator-Activated Receptor Gamma
    Gene Symbol PPARG
    Protein Tag GST Tag, His Tag
    Validated Application Assay Standard
    Protein Family Other proteins
    Expression System Insect
    Product Type PPARG Recombinant Human Protein
    For Use With (Application) Analysis of protein stucture and used in cell biology research applications
    Format Lyophilized
    Gene ID (Entrez) 5468

    Expressed from DNA sequence encoding human PPARG isoform 2 (Met 1-Tyr 505) fused to N-terminal polyhistidine-tagged GST tag at N-Terminus

    • Formulation: 50mM Tris, 100mM NaCl, 20% Glycerol, 0.3 mM DTT, pH 7.4

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit