PPD-2 DPD Powder Pop™ Dispensers for Chlorine Testing
Use with any DPD-based photometer or test kit
Manufacturer: HF SCIENTIFIC INC 10447A
 View more versions of this product
Catalog No. 15-393-68
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) For free chlorine testing
    For Use With (Equipment) For Hach™ and Hanna photometers
    Volume (English) 0.33 oz.
    Volume (Metric) 10mL
    Product Type Chlorine Reagent Powder Pop Dispenser
    Quantity 400 Tests
  • Description & Specifications
Related Products
Product Content Feedback