Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Fully Adjustable PPE Holder

Poltex Fully Adjustable PPE Holder holds virtually any boxed PPE item and adapts to changing products

Manufacturer:  Poltex FULADJST4M

 View more versions of this product

Catalog No. 22-000-277


Add to cart

Description

Description

  • Easy adjustment of width and depth for when PPE size changes from manufacturer
  • Super durable 0.25in thick PETG construction
  • Powerful magnets or keyhole slots for mounting
Specifications

Specifications

Clear
4in.
1
Adjusts from 3.1 to 5.3in.
Magnet
Adjusts from 6.6 to 11.5in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit