Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPEF2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548692

Catalog No. PA548692


Add to cart

Description

Description

This gene encodes a member of the serine/threonine protein phosphatase with EF-hand motif family. The protein contains a protein phosphatase catalytic domain, and at least two EF-hand calcium-binding motifs in its C terminus. Although its substrate(s) is unknown, the encoded protein, which is expressed specifically in photoreceptors and the pineal, has been suggested to play a role in the visual system. This gene shares high sequence similarity with the Drosophila retinal degeneration C gene.
Specifications

Specifications

PPEF2
Polyclonal
PBS with 0.09% sodium azide
O14830
Serine/threonine-protein phosphatase with EF-hands 2, PPEF-2, PPEF2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5470
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PPEF2
Liquid
PPEF2
KLH conjugated synthetic peptide between 304-333 amino acids from the Central region of human PPEF2.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit