Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPFIBP1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP156315
Description
PPFIBP1 Polyclonal specifically detects PPFIBP1 in Human samples. It is validated for Western Blot.Specifications
PPFIBP1 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Q86W92-5 | |
PPFIBP1 | |
Synthetic peptides corresponding to PPFIBP1(PTPRF interacting protein, binding protein 1 (liprin beta 1)) The peptide sequence was selected from the N terminal of PPFIBP1. Peptide sequence PFMGSLRALHLVEDLRGLLEMMETDEKEGLRCQIPDSTAETLVEWLQSQM | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 100%; Chicken: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Canine: 92%; Guinea pig: 92%. | |
Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
Purified |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
hSGT2liprin related protein, hSgt2p, KIAA1230, L2, liprin-beta 1, liprin-beta-1, Protein tyrosine phosphatase receptor type f polypeptide-interactingprotein-binding protein 1, protein-tyrosine phosphatase receptor-type f polypeptide-interactingprotein-binding protein 1, PTPRF interacting protein, binding protein 1 (liprin beta 1), PTPRF-interacting protein-binding protein 1, SGT2 | |
Rabbit | |
Protein A purified | |
RUO | |
8496 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title