Learn More
Aviva Systems Biology Corp PPIA Antibody - N-terminal region (ARP51353_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP51353_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPIA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PPIA. Alternate names: CYPA, CYPH, MGC117158, MGC12404, MGC23397ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast
Catalog No. 50-014-60557