Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPIAL4A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549103

Catalog No. PA549103


Add to cart

Description

Description

PPIases accelerate the folding of proteins. It catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.
Specifications

Specifications

PPIAL4A
Polyclonal
PBS with 0.09% sodium azide
Q9Y536
Peptidyl-prolyl cis-trans isomerase A-like 4A/B/C, PPIase A-like 4A, Chromosome 1-amplified sequence 2, COAS-2, Cyclophilin homolog overexpressed in liver cancer, PPIAL4A, COAS2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
653505
Western Blot
Unconjugated
PPIAL4A
Liquid
PPIAL4A
KLH conjugated synthetic peptide between 66-92 amino acids from the Central region of human PPIAL4A.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit