Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
PPIH Mouse anti-Human, Clone: OTI1C6, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF803474
Catalog No. 50-166-6519
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. This protein is a specific component of the complex that includes pre-mRNA processing factors PRPF3, PRPF4, and PRPF18, as well as U4/U5/U6 tri-snRNP. This protein has been shown to possess PPIase activity and may act as a protein chaperone that mediates the interactions between different proteins inside the spliceosome.
Specifications
Specifications
PPIH
Monoclonal
Unconjugated
PPIH
Lyophilized
PPIH
Full length human recombit protein of human PPIH produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI1C6
PBS with 8% trehalose and no preservative; pH 7.3
O43447
CYP-20, CYPH, SnuCyp-20, USA-CYP
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
10465