Learn More
PPIL1 Mouse anti-Human, Clone: OTI2D1, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF803692
Catalog No. 50-166-6631
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is a member of the cyclophilin family of peptidylprolyl isomerases (PPIases). The cyclophilins are a highly conserved, ubiquitous family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV-1 virions. Based on similarity to other PPIases, this protein could accelerate the folding of proteins and might catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.
Specifications
Specifications
PPIL1
Monoclonal
Unconjugated
PPIL1
Lyophilized
PPIL1
Full length human recombit protein of human PPIL1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI2D1
PBS with 8% trehalose and no preservative; pH 7.3
Q9Y3C6
CGI-124, CYPL1, PPIase, hCyPX
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
51645