Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPIL4 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549056

Catalog No. PA549056

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene is a member of the cyclophilin family of peptidylprolyl isomerases. The cyclophilins are a highly conserved family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV-1 virions.
Specifications

Specifications

PPIL4
Polyclonal
PBS with 0.09% sodium azide
Q9CXG3
Peptidyl-prolyl cis-trans isomerase-like 4, PPIase, Cyclophilin-like protein PPIL4, Rotamase PPIL4, PPIL4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
67418
Western Blot
Unconjugated
Ppil4
Liquid
PPIL4
KLH conjugated synthetic peptide between 122-151 amino acids from the N-terminal region of human PPIL4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit