Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRR1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548954

Catalog No. PA548954

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with monkey based on sequence homology.

May negatively regulate the 4-1BB-mediated signaling cascades which result in the activation of NK-kappaB and JNK1. Probable substrate recognition subunit of an ECS (Elongin BC- CUL2/5-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
Specifications

Specifications

LRR1
Polyclonal
PBS with 0.09% sodium azide
Q96L50
Leucine-rich repeat protein 1, 4-1BB-mediated-signaling molecule, 4-1BBlrr, LRR-repeat protein 1, LRR-1, Peptidylprolyl isomerase-like 5, LRR1, PPIL5
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
122769
Western Blot
Unconjugated
LRR1
Liquid
LRR1
KLH conjugated synthetic peptide between 127-153 amino acids from the Central region of human PPIL5.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit