Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPIL6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548904

Catalog No. PA548904

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PPIases accelerate the folding of proteins. It catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.
Specifications

Specifications

PPIL6
Polyclonal
PBS with 0.09% sodium azide
Q8IXY8
Peptidyl-prolyl cis-trans isomerase-like 6, PPIase, Cyclophilin-like protein PPIL6, Rotamase PPIL6, PPIL6
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
285755
Western Blot
Unconjugated
PPIL6
Liquid
PPIL6
KLH conjugated synthetic peptide between 30-58 amino acids from the N-terminal region of human PPIL6.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit