Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPM1D Phosphatase Inhibitor

Acts as a PPM1D inhibitor

$104.90 - $400.00

Specifications

CAS 176957-55-4
Formula Weight 272.40g/mol
Synonym 2,5-Bis-(2-Thienylidene)cyclopentanone; CCT007093
Storage Requirements 2° to 8°C
Format Solid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ64165MA View Documents Alfa Aesar™
J64165MA
10mg Each for $104.90
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ64165MF View Documents Alfa Aesar™
J64165MF
50mg Each for $400.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

176957-55-4
2,5-Bis-(2-Thienylidene)cyclopentanone; CCT007093
Solid
Acts as a PPM1D inhibitor
Phosphatase Inhibitor
272.40g/mol
2° to 8°C
Air Sensitive
Yellow
Safety and Handling

Safety and Handling

Recommended Storage : Keep cold; Store under Argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit