Learn More
Alfa Aesar™ PPM1D Phosphatase Inhibitor
Acts as a PPM1D inhibitor
Manufacturer:  Alfa Aesar™ J64165MA
 View more versions of this product
Catalog No. AAJ64165MA
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
Acts as a PPM1D inhibitor
Solid
2° to 8°C
176957-55-4
272.40g/mol
Light Sensitive
Yellow
Phosphatase Inhibitor
10mg
C15H12OS2
2,5-Bis-(2-Thienylidene)cyclopentanone; CCT007093