Learn More
Aviva Systems Biology Corp PPM1K Antibody - middle region (ARP53031_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53031_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPM1K. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PPM1K. Alternate names: DKFZp667B084, DKFZp761G058, PP2Cm, PTMP, UG0882E07, hPTMP, PP2CkappaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60897