Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPM1M Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$152.22 - $436.00
Specifications
Antigen | PPM1M |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15663020
|
Novus Biologicals
NBP15663020UL |
20 μL |
Each for $152.22
|
|
NBP156630
|
Novus Biologicals
NBP156630 |
100 μL |
Each for $436.00
|
|
Description
PPM1M Polyclonal specifically detects PPM1M in Human samples. It is validated for Western Blot.Specifications
PPM1M | |
Polyclonal | |
Rabbit | |
Protein Phosphatase | |
Q96MI6 | |
132160 | |
Synthetic peptides corresponding to PPM1M (protein phosphatase, Mg2+/Mn2+ dependent, 1M) The peptide sequence was selected from the middle region of PPM1M. Peptide sequence VYPELLAGEFTRLEFPRRLKGDDLGQKVLFRDHHMSGWSYKRVEKSDLKY. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
FLJ32332, PP2CEEC 3.1.3.16, PP2Ceta, PP2C-eta, PPM1E, protein phosphatase 1M, protein phosphatase 1M (PP2C domain containing), protein phosphatase 2C eta, protein phosphatase 2C eta-2, Protein phosphatase 2C isoform eta, protein phosphatase, Mg2+/Mn2+ dependent, 1M | |
PPM1M | |
IgG | |
Affinity Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title