Learn More
Aviva Systems Biology Corp PPME1 Antibody - N-terminal region (ARP56844_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56844_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPME1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PPME1. Alternate names: FLJ22226, PME-1ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Rat
Catalog No. 50-014-62008