Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPME1 Mouse anti-Human, Clone: 29G3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259487100UL

 View more versions of this product

Catalog No. NBP259487X


Add to cart

Description

Description

PPME1 Monoclonal antibody specifically detects PPME1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

PPME1
29G3
Unconjugated
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
EC 3.1.1, EC 3.1.1.-, FLJ22226, PME-1PME1, protein phosphatase methylesterase 1, protein phosphatase methylesterase-1
Mouse
IgG1 Kappa
100 ul
Lipid and Metabolism
Primary
51400
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
1 mg/mL
Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100
Purified
PPME1
Recombinant human PPME1 (1-386aa) purified from E. coli
Protein A purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only