Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ppNPY Guinea Pig anti-Mouse, Rat, Polyclonal, Neuromics™ Non-distribution product as customer accommodation.

Guinea Pig Polyclonal Antibody

$245.52 - $579.50

Specifications

Antigen ppNPY
Concentration 1 mg/mL
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-0015 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
GP14107150UL
150 μL
Each for $579.50
Add to cart
 
50-210-0016 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
GP1410750UL
50 μL
Each for $245.52
Add to cart
 
Description

Description

ppNPY Polyclonal antibody specifically detects ppNPY in Mouse, Rat samples. It is validated for Immunohistochemistry
Specifications

Specifications

ppNPY
Immunohistochemistry
Unconjugated
Guinea Pig
Mouse, Rat
P07808, P57774
100912228, 109648, 24604
IgG
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
NPY
1 mg/mL
Polyclonal
Liquid, Purified
RUO
whole serum with 0.05% sodium azide
Neuropeptide-Y
SDLLMRESTENAPRTR
Polyclonal
Protein G
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit