Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPOX Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550733

Catalog No. PA550733


Add to cart

Description

Description

The antibody detects endogenous levels of total PPOX protein.

This gene encodes the penultimate enzyme of heme biosynthesis, which catalyzes the 6-electron oxidation of protoporphyrinogen IX to form protoporphyrin IX. Mutations in this gene cause variegate porphyria, an autosomal domit disorder of heme metabolism resulting from a deficiency in protoporphyrinogen oxidase, an enzyme located on the inner mitochondrial membrane. Alternatively spliced transcript variants encoding the same protein have been identified.
Specifications

Specifications

PPOX
Polyclonal
Unconjugated
PPOX
Liquid
PPOX
Synthetic peptide corresponding to residues near the C terminal of human protoporphyrinogen oxidase
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
2.5 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
P50336, P51175
VP, PPO, V290M
Rabbit
IgG
100 μL
-20°C
Primary
19044, 5498
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit