Learn More
PPP1R10 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 244501AP
Catalog No. 50-173-4366
Add to cart
Description
Description
RNA polymerases transcribe nuclear genes for ribosomal RNA, thus representing ribosomal biogenesis. RNA polymerase I (Pol I) is located in the nucleolus and transcribes class I genes, which code for large ribosomal RNA. Different subunits of the Pol I transcription machinery are targets of various physiological stimuli, which suggests that multiple signaling pathways are involved in carrying out Pol I transcription. RPA40 and RPA16 are subunits of Pol I that associate with each other at an early stage of RNA polymerase I assembly. RPA40 is essential for the function and integrity of the complex and is also an essential subunit of RNA polymerase III (Pol III). RPA40, RPA16 and RPA135 encode the three subunits of RNA polymerase I, respectively. RPA194 is the largest subunit of RNA Pol I and is not a component of Pol II and Pol III.
Specifications
Specifications
PPP1R10
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96QC0
CAT53, FB19, p99, PNUTS, PP1 binding protein of 114 kDa, PP1R10, PPP1R10, Protein FB19
Rabbit
IgG
150 μL
-20°C
Primary
5514
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
PPP1R10
Liquid
PPP1R10
PPP1R10 Fusion Protein Ag20091
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human