Learn More
PPP1R14B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 184761AP
Catalog No. 50-173-2163
Add to cart
Description
Description
This gene is thought to play a role in a complex developmental pathway and the regulation of genes involved in cerebellar development. Mutations in this gene have been associated with autosomal recessive spinocerebellar ataxia.
Specifications
Specifications
PPP1R14B
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q62084, Q8K3F3, Q96C90
PHI 1, PLCB3N, PNG, PPP1R14B, SOM172
Rabbit
IgG
150 μL
-20°C
Primary
18938, 259225, 26472
Western Blot
Unconjugated
PPP1R14B
Liquid
Ppp1r14b
PPP1R14B Fusion Protein Ag13373
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat