Learn More
PPP1R15A Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI3D12, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA504310S
Catalog No. 50-167-5031
Add to cart
Description
Description
This gene is a member of a group of genes whose transcript levels are increased following stressful growth arrest conditions and treatment with DNA-damaging agents. The induction of this gene by ionizing radiation occurs in certain cell lines regardless of p53 status, and its protein response is correlated with apoptosis following ionizing radiation.
Specifications
Specifications
PPP1R15A
Monoclonal
0.25 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O75807, P17564, Q6IN02
GADD34
Mouse
IgG2b
30 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
171071, 17872, 23645, 476419
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI3D12
Unconjugated
PPP1R15A
Liquid
PPP1R15A
Full length human recombit protein of human PPP1R15A produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Mouse, Rat