Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1R3B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548980

Catalog No. PA548980

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Acts as a glycogen-targeting subunit for phosphatase PP1. Facilitates interaction of the PP1 with enzymes of the glycogen metabolism and regulates its activity. Suppresses the rate at which PP1 dephosphorylates (inactivates) glycogen phosphorylase and enhances the rate at which it activates glycogen synthase and therefore limits glycogen breakdown. Its activity is inhibited by PYGL, resulting in inhibition of the glycogen synthase and glycogen phosphorylase phosphatase activities of PP1. Dramatically increases basal and insulin-stimulated glycogen synthesis upon overexpression in hepatocytes.
Specifications

Specifications

PPP1R3B
Polyclonal
PBS with 0.09% sodium azide
Q86XI6
Protein phosphatase 1 regulatory subunit 3B, Hepatic glycogen-targeting protein phosphatase 1 regulatory subunit GL, Protein phosphatase 1 regulatory subunit 4, PP1 subunit R4, Protein phosphatase 1 subunit GL, PTG, PPP1R3B, PPP1R4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79660
Western Blot
Unconjugated
PPP1R3B
Liquid
PPP1R3B
KLH conjugated synthetic peptide between 205-231 amino acids from the C-terminal region of human PPP1R3B.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit