Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP1R3E Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572336
Description
Acts as a glycogen-targeting subunit for PP1. PP1 is involved in glycogen metabolism and contributes to the activation of glycogen synthase leading to an increase in glycogen synthesis.
Specifications
PPP1R3E | |
Polyclonal | |
Unconjugated | |
PPP1R3E | |
Protein phosphatase 1 regulatory subunit 3E; PPP1R3E | |
Rabbit | |
Protein A | |
RUO | |
90673 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.52 mg/mL | |
PBS with 0.09% sodium azide | |
Q9H7J1 | |
PPP1R3E | |
KLH conjugated synthetic peptide between 159-192 amino acids from the Central region of human PPP1R3E | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction