Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1R3F Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548620

Catalog No. PA548620

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Glycogen-targeting subunit for protein phosphatase 1 (PP1).
Specifications

Specifications

PPP1R3F
Polyclonal
PBS with 0.09% sodium azide
Q6ZSY5
Protein phosphatase 1 regulatory subunit 3F, R3F, PPP1R3F
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
89801
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PPP1R3F
Liquid
PPP1R3F
KLH conjugated synthetic peptide between 549-578 amino acids from the C-terminal region of human PPP1R3F.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit