Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPP1R3G Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563295
Description
Immunogen sequence: DAKKEPGAEC FHFSLCLPPG LQPEDEEDAD ERGVAVHFAV CYRCAQGEYW DNNAGANYTL RY Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 77%.
Glycogen-targeting subunit for protein phosphatase 1 (PP1). Involved in the regulation of hepatic glycogenesis in a manner coupled to the fasting-feeding cycle and distinct from other glycogen-targeting subunits.
Specifications
PPP1R3G | |
Polyclonal | |
Unconjugated | |
PPP1R3G | |
PPP1R3G; Protein phosphatase 1 regulatory subunit 3G; protein phosphatase 1, regulatory (inhibitor) subunit 3G; protein phosphatase 1, regulatory subunit 3G; putative protein phosphatase 1 regulatory inhibitor subunit 3G | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
648791 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
B7ZBB8 | |
PPP1R3G | |
Recombinant protein corresponding to Human PPP1R3G. Recombinant protein control fragment (Product #RP-100689). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction