Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1R8 Mouse anti-Human, Clone: OTI4D5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526694

Catalog No. PIMA526694


Add to cart

Description

Description

This gene, through alternative splicing, encodes three different isoforms. Two of the protein isoforms encoded by this gene are specific inhibitors of type 1 serine/threonine protein phosphatases and can bind but not cleave RNA. The third protein isoform lacks the phosphatase inhibitory function but is a single-strand endoribonuclease comparable to RNase E of E. coli. This isoform requires magnesium for its function and cleaves specific sites in A+U-rich regions of RNA.
Specifications

Specifications

PPP1R8
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q12972
PPP1R8; NIPP1; protein phosphatase 1, regulatory (inhibitor) subunit 8; ARD1; NIPP-1; PRO2047; ard-1; activator of RNA decay; nuclear subunit of PP-1; homolog of E.coli RNase E; nuclear inhibitor of protein phosphatase-1; protein phosphatase-1, regulatory subunit-8; ARD-1; RNase E; protein phosphatase 1 regulatory inhibitor subunit 8; protein phosphatase 1 regulatory subunit 8
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5511
Immunohistochemistry (Paraffin), Western Blot
OTI4D5
Unconjugated
PPP1R8
Liquid
PPP1R8
Human recombit protein fragment corresponding to amino acids 1-225 of PPP1R8 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit