Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP2R1A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP219907
Description
PPP2R1A Polyclonal specifically detects PPP2R1A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
PPP2R1A | |
Polyclonal | |
Unconjugated | |
PBS, 20% Glycerol (pH7) with 0.025% Proclin 300 | |
Medium tumor antigen-associated 61 kDa protein, MGC786, PP2A subunit A isoform PR65-alpha, PP2A subunit A isoform R1-alpha, PP2A-Aalpha, PP2AAALPHA, PR65A, protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), alphaisoform, protein phosphatase 2 (formerly 2A), regulatory subunit A, alpha isoform, protein phosphatase 2, regulatory subunit A, alpha, serine/threonine protein phosphatase 2A, 65 kDa regulatory subunit A, alphaisoform, serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A alphaisoform | |
Rabbit | |
65 kDa | |
0.1 mL | |
Protein Phosphatase | |
5518 | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.96mg/mL | |
Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:2500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation, Immunohistochemistry-Paraffin 1:100-1:2500 | |
P30153 | |
PPP2R1A | |
Recombinant protein encompassing a sequence within the center region of human PPP2R1A. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse. Species reactivity based on sequence identity: Pig (100%), Rat (100%), Rhesus Monkey (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction