Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP2R1B Mouse anti-Human, Mouse, Rat, Clone: 1496CT356.164.25.226, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524735

Catalog No. PIMA524735


Add to cart

Description

Description

In eukaryotes, the phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions, including division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the protein phosphatases. In general, the protein phosphatase (PP) holoenzyme is a trimeric complex composed of a regulatory subunit, a variable subunit and a catalytic subunit. Four major families of protein phosphatase catalytic subunits have been identified, designated PP1, PP2A, PP2B (calcineurin) and PP2C. An additional protein phosphatase catalytic subunit, PPX (also known as PP4), is a putative member of a novel PP family. The PP2A family comprises subfamily members PP2A alpha and PP2A beta. The PP2A catalytic subunit associates with a variety of regulatory subunits. Regulatory subunits include PP2A-A alpha and -A beta, PP2A-B alpha and -B beta, PP2A-C alpha and -C beta, and PP2A-B56 alpha and -B56 beta.
Specifications

Specifications

PPP2R1B
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide
P30154, Q4QQT4, Q7TNP2
Serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A beta isoform, PP2A subunit A isoform PR65-beta, PP2A subunit A isoform R1-beta, PPP2R1B
Mouse
IgG1, kappa
400 μL
-20°C
Primary
315648, 5519, 73699
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1496CT356.164.25.226
Unconjugated
PPP2R1B
Liquid
PPP2R1B
human PPP2R1B recombit protein
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit