Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP2R2A Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen PPP2R2A
Applications Immunofluorescence, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093791
View Documents
Novus Biologicals
NBP304724100UL
100 μg Each for $320.71
Add to cart
 
NB093792
View Documents
Novus Biologicals
NBP30472420UL
20 μg Each for $138.37
Add to cart
 
Description

Description

PPP2R2A Polyclonal antibody specifically detects PPP2R2A in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

PPP2R2A
Unconjugated
Rabbit
Human, Mouse
B55A, B55ALPHA, DKFZp686N05117, FLJ26613, FLJ41727, MGC52248, PP2A subunit B isoform alpha, PP2A subunit B isoform B55-alpha, PP2A subunit B isoform PR55-alpha, PP2A subunit B isoform R2-alpha, PR52A, PR55A, protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), alphaisoform, protein phosphatase 2 (formerly 2A), regulatory subunit B (PR52), alpha isoform, protein phosphatase 2 (formerly 2A), regulatory subunit B, alpha isoform, protein phosphatase 2, regulatory subunit B, alpha, serine/threonine protein phosphatase 2A, 55 KDA regulatory subunit B, alphaisoform, serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B alphaisoform
Recombinant fusion protein containing a sequence corresponding to amino acids 1-447 of human PPP2R2A (NP_002708.1).
Polyclonal
Affinity purified
Immunofluorescence, Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
5520
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit