Learn More
Aviva Systems Biology Corp PPP2R3A Antibody - N-terminal region (ARP56410_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56410_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP2R3A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PPP2R3A. Alternate names: PPP2R3, PR130, PR72ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61848